Anti-CFTR Mouse Monoclonal Antibody [clone: CF3]
Katalogové číslo NOVUNB300-511
Dodavatel: Novus Biologicals
New Product
Specifikace
- Druh protilátky:Primární
- Název antigenu:Cystic Fibrosis Transmembrane Regulator
- Symbol antigenu:CFTR
- Klonovalita:Monoclonal
- Klon:CF3
- Spojení:Unconjugated
- Průtoková cytometrie:Yes
- Hostitel:Mouse
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Imunoprecipitace:Yes
- Izotyp:IgM
- Reaktivita:Chicken,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Neutralizace:Yes
- Identifikační číslo genu:1080
- Synonyma antigenu:dJ760C5.1|ABC35|MRP7EC 3.6.3.49|ATP-binding cassette(sub-family C|Channel conductance-controlling ATPase|ATP-binding cassette sub-family C member 7|cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C|ABCC7CF|cAMP-dependent chloride channel|EC 3.6.3|TNR-CFTR|ATP-binding cassette transporter sub-family C member 7|member 7)|CFTR/MRP|cystic fibrosis transmembrane conductance regulator
- Skladovací pufr:Diluted ascites
- Skladovací teplota:Store at −20 °C. Avoid freeze-thaw cycles.
- Imunogen:Synthetic Peptide: G(103) R I I A S Y D P D N K E E R(117)
- Čištění:Unpurified
- Bal.:100 µl
Specifikace
O tomto produktu
The CFTR Antibody (CF3) from Novus Biologicals is a mouse monoclonal antibody to CFTR. This antibody reacts with human, mouse, rat, chicken. The CFTR Antibody (CF3) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Blocking/Neutralising.
Type: Primary
Antigen: CFTR
Clonality: Monoclonal
Clone: CF3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat, Chicken