Order Entry
Czech Republic
ContactUsLinkComponent
Trichoderma viride cellulase
Trichoderma viride cellulase
Supplier:  Merck
Certificates
Trichoderma viride cellulase
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    9012-54-8
  • Species:
    Trichoderma viride
  • MDL No.:
    MFCD00081510
  • Enzyme name:
    Cellulase

Specifications

Related Information

Frequently Bought Together