3,7-Dihydroxy-2-naphthoic acid, Sigma-Aldrich®
Catalog # SIAL275271-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:204.18 g/mol
:M-Clarity™ quality level = MQ200
CAS Number:83511-07-3
Formula:C₁₁H₈O₄
MDL Number:MFCD00010253