3,5-Dihydroxy-2-naphthoic acid, Sigma-Aldrich®
Catalog # SIAL275298-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:204.18 g/mol
:M-Clarity™ quality level = MQ200
EINECS:201-900-8
CAS Number:89-35-0
Formula:C₁₁H₈O₄