(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:82473-24-3
:
Formula:C₃₂H₅₈N₂O₈S
:630.89 g/mol
Melting Pt:184…186 °C
:M-Clarity™ quality level = MQ200
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together