ContactUsLinkComponent
Bordet Gengou Agar Base, Dehydrated Culture Media
  HIMEM175-500G
 :  HIMEDIA
New Product
 :  
Bordet Gengou Agar Base, Dehydrated Culture Media
  HIMEM175-500G
 :  HIMEDIA
New Product
 :  
  • Pk:
    1
  • Description:
    Bordet Gengou agar base, 500 g
For the detection and isolation of Bordetella pertussis and Bordetella parapertussis.
  • Microbiological culture media
  • Packaged in 500 g poly bottles