
N,N-Diethyl-p-phenylenediammonium sulphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6283-63-2
:
Formula:C₁₀H₁₈N₂O₄S
MDL Number:MFCD00012993
:262.33 g/mol
Boiling Pt:274…275 °C (1013 hPa)
Melting Pt:184…186 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:228-500-6
:GHS07
Frequently Bought Together