Lentiviral Control Particles
Catalog # ORIGTR30021V5
Supplier: OriGene
CAS Number:
Specifications
- Pk:1 KIT
- Description:Control Lenti particles, shRNA scramble, expressing GFP and puro, 5×0,5 ml
- Seq:01
Specifications
About this item
Lenti particles carrying a 29-mer scrambled sequence cassette and serve as a negative control.
- Prepackaged shRNA transduction control (GFP) scramble particles, ready for transduction
- Puro or blasticidin selection marker driven by SV40
- GFP or RFP as a reporter
Applications: Monitor transduction efficiency, MOI optimisation, non-target control.