Order Entry
Czech Republic
ContactUsLinkComponent
 
Disodium naphthalene-1,5-disulphonate, Sigma-Aldrich®
Catalog # SIAL70240-1KG
Supplier:  Merck
CAS Number:  
Disodium naphthalene-1,5-disulphonate, Sigma-Aldrich®
Catalog # SIAL70240-1KG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :332.27 g/mol
EINECS:216-732-0
CAS Number:1655-29-4
Formula:C₁₀H₆Na₂O₆S₂
MDL Number:MFCD00064178