ContactUsLinkComponent
Magnetic Stirrer with Stand
Magnetic Stirrer with Stand
Catalog # METR28010040
Supplier:  METROHM
New Product
Magnetic Stirrer with Stand
Catalog # METR28010040
Supplier:  METROHM
New Product
Specifications
  • Description:
    Magnetic stirrer with stand
  • Pk:
    1
Specifications
About this item
Magnetic stirrer including base plate, support rod and electrode holder for supplementing the Titrino plus, Dosimat plus, titrandos, sample processors, 805 Dosimat and 780/781 pH meters as well as the 856 and 867 measuring modules.
With permanently attached cable for MSB (Metrohm Serial Bus).