Order Entry
Czech Republic
ContactUsLinkComponent
Screw Caps with Dropping Pipette for Thread Bottles
Screw Caps with Dropping Pipette for Thread Bottles
Catalog # MARI5459903
Supplier:  MARIENFELD
New Product
Screw Caps with Dropping Pipette for Thread Bottles
Catalog # MARI5459903
Supplier:  MARIENFELD
New Product

Specifications

  • Thread:
    GL 18
  • Description:
    Screw cap with 78 mm dropping pipette
  • Material:
    PP
  • Pk:
    100
  • For bottles:
    30 ml thread bottles ml

Specifications

About this item

Screw caps with dropping pipette.

  • Made of white polypropylene
  • With white rubber teat
  • With dropping pipette with bent, spherical tip made of clear glass

Screw caps GL 18 for thread bottles.

Packaging: Minimum order quantity: 100 pieces each type.