ContactUsLinkComponent
Dispenser, 7 Circles ProLine
Dispenser, 7 Circles ProLine
Catalog # EVONPROB01HCMC
Supplier:  Deb-STOKO
New Product
Dispenser, 7 Circles ProLine
Catalog # EVONPROB01HCMC
Supplier:  Deb-STOKO
New Product
Specifications
  • Capacity:
    1000 ml
  • Description:
    7 Circles ProLine dispenser
  • Pk:
    1
Specifications
About this item
For exclusive use with SC Johnson (Deb) skincare products to create tailored skincare and hand hygiene programs. The 7 Circles ProLine Dispenser is designed for use with Cutan 1 litre cartridges.
  • Easy visual identification of product type to encourage correct product usage and compliance
  • Simple to understand and language neutral to show best practice method of use
  • Ideal for multinational workforces
  • Large push buttons minimise the operation force required to allow easy use by hand or elbow
  • Suitable in hygiene sensitive areas
  • Fixed product dose delivers the exact amount required, controlling usage and reducing waste
  • Unique locking mechanism helps prevent unauthorised access, theft, and product contamination
  • Sight glass provides product level visibility in the cartridge to help determine when replacement is necessary
  • Air-tight cartridges are designed to collapse to minimise product waste
All Deb dispensers are manufactured from durable plastic components and rigorously tested for high usage environments, providing assurance of quality and longevity. BioCote antimicrobial protection technology is incorporated into the plastic buttons of all Deb manually operated, wall-mounted skincare dispensers. The button is the primary contact point for people using these dispensers and BioCote technology provides life-long antimicrobial protection to inhibit the growth of a broad spectrum of bacteria and moulds that could potentially cause illness.
Designed for exclusive use with Deb skincare system products to enable cartridges to be replaced in seconds, reducing maintenance time and costs. The ultrasonically sealed, airless cartridge and pump mechanisms prevent the ingress of air into the cartridge during use to prevent product contamination and ensure ‘fresh’ product is dispensed every time.
Ordering information: Cartridges sold separately.