Anios Oxyfloor Detergent Disinfectant and Cleaner
SHIE3101330
New Product
- Capacity:1 kg
- Description:Disinfectant and cleaning agent for floors and surfaces
- Pk:4
This broad-spectrum disinfectant and cleaner is great for all types of floors and surfaces, such as work surfaces, bedside tables, handrails, railings and many more.
- Odorless powder - white powder with low granulometry releases peracetic acid (keep the hydroscopic powder dry)
- Double effect - disinfection and cleaning in one
- Excellent alternative to chlorine preparations
- Simple dosing - no danger of overdosing
- pH after dissolution (0,5 to 2%) in drinking water (minimum 8 to maximum 10 minutes) - a blue-coloured solution is formed, which can be used for 8 hours
- Composition without perborate