
Zincate electroless immersion zinc plating solution
Catalog # SIAL901661-1L
Supplier: Merck
New Product
CAS Number:
Specification Test Results
- Appearance (Colour):Colourless to Light Yellow
- Appearance (Form):Liquid
- pH:≥10
- Recommended Retest Period:15 Months