Order Entry
Czech Republic
ContactUsLinkComponent
 
Mueller Hinton Fastidious Agar
Catalog # LIOF10132
Supplier:  LIOFILCHEM
New Product
CAS Number:  
Mueller Hinton Fastidious Agar
Catalog # LIOF10132
Supplier:  LIOFILCHEM
New Product
CAS Number:  

Specifications

  • Pk:
    20
  • Description:
    Mueller Hinton Fastidious Agar (Horse blood 5% + 20 mg/L β-NAD)
  • Storage temperature:
    2…8 °C
  • Formulation:
    Meat extract: 2,0 g/ml, Casamino acids, Technical: 17,5 g/ml, Starch: 1,5 g/ml, β-NAD: 0,02 g/ml, Agar: 15,0 g/ml, Horse defibrinated blood: 50 ml
  • Incubation:
    35...39 °C for 48 hours
  • pH:
    7,3 ±0,1
  • Clinical:
    +

Specifications

About this item

Mueller Hinton fastidious agar (Horse blood 5% + 20 mg/L β-NAD) is recommended by EUCAST for antimicrobial susceptibility testing of Streptococcus spp (including S. pneumoniae), Haemophilus spp and several other fastidious organisms.