CrossLab CS Gas Leak Detector
Catalog # HEWLG6693A
Supplier: Agilent
Specifications
- Description:Handheld electronic gas leak detector
- Pk:1
Specifications
About this item
Leak detector will detect leaks or verify leak-free tubing and fittings for various gas types, including hydrogen, nitrogen, helium, and others, throughout the lab or site.
- Unique design
- Used for other analytical techniques like LC/MS and ICP-MS
The cartridge design combines the two most critical GC flow path monitoring tasks, including flow rate and leak detection monitoring, into a single handheld system with the bundle offering.