VWR® Reagent Reservoirs
Supplier: VWR Collection
New Product
Certificates
About this item
Reagent reservoirs are designed for efficient liquid transfer, making them an ideal choice for repetitive pipetting tasks. These reagent reservoirs for multichannel pipettes provide a trough wide enough to fit 8 and 12 channel pipettes.
- Trough design with graduations on the inside wall and pour-off spouts on four corners
- Slightly tilted inner surface helps reduce residue
- Non sterile or sterilised options available, achieving SAL 10⁻⁶ (ISO 11137)
- Reservoirs are lot-certified, non-pyrogenic, DNase- and RNase-free
VWR® reagent reservoirs are an excellent choice for holding different reagents, including those used in the setup of high-throughput screening, ELISA, PCR, and cell culture experiments.
The 25 ml reservoirs are available with a single well or 3-well design (each holding 5 ml). The 50 ml reservoirs are available in PVC, PET, and PS.
Certifications: ISO 13485 and ISO 9001.
Packaging: Available with inner packaging formats of 1/bag, 5/bag, and bulk.
Specifications
- Catalog No:
- 613-7299
- 613-7310
- 613-7289
- 613-7311
- 613-7290
- 613-7301
- 613-7312
- 613-7291
- 613-7287
- 613-7306
- 613-7307
- 613-7297
- 613-7308
- 613-7284
- 613-7314
- 613-7285
- 613-7293
- 613-7304
- 613-7286
- 613-7294
- 613-7305
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Frequently Bought Together