Peristaltic Pump Tubing and Consumable Kit, Pharm-A-line
Supplier: METROHM
New Product
Certificates
About this item
The Pharm-A-line pump tubing is used for the transport of both aqueous and nonaqueous solutions and must be operated in connection with the white press clamp in the pump module. Two pump tubings each and associated press clamps for replacement in the OMNIS pump module. One consumable kit can be used for re-equipping two peristaltic pumps of the pump module.
Frequently Bought Together