ContactUsLinkComponent
TASKI Sani Calc Pur-Eco W3b Washroom Descaler
TASKI Sani Calc Pur-Eco W3b Washroom Descaler
Catalog # DIVY7515243
Supplier:  Diversey
New Product
TASKI Sani Calc Pur-Eco W3b Washroom Descaler
Catalog # DIVY7515243
Supplier:  Diversey
New Product
Specifications
  • Capacity:
    6 L
  • Description:
    Washroom descaler, slightly perfumed
  • Pk:
    6 Bottle
Specifications
About this item
Powerful limescale remover for acid resistant hard surfaces.
  • Fast acting acidic, based on citric acid formula
  • Excellent cleaning performance
  • Strong descaling properties
  • No scrubbing necessary at high concentration
Certifications: EU Ecolabel (EU Flower).