Specifications
- Pk:500 mL
- Source:
- Species:
- Description:Endothelial cell growth medium MV2
- Environmentally Preferable:
Specifications
About this item
Endothelial cell growth medium MV2 was developed for the in vitro cultivation of endothelial cells from microvascular vessels, the coronary artery, and the aorta.
- Form: Aqueous solution
- Shipped in: Wet ice
- Techniques: Cell culture; mammalian: suitable
Application: The media are optimised for primary human cells but can also be used for bovine, porcine, murine, rat and dog endothelial cells from microvascular vessels as well as for various endothelial cell lines. Recommended for: Human Dermal Microvascular Endothelial Cells (HDMEC), Human Coronary Artery Endothelial Cells (HCAEC), Human Aortic Endothelial Cells (HAoEC), Human Pulmonary Microvascular Endothelial Cells (HPMEC), Human Dermal Lymphatic Endothelial Cells (HDLEC), Human Dermal Blood Endothelial Cells (HDBEC), Human Cardiac Microvascular Endothelial Cells (HCMEC), Human Uterine Microvascular Endothelial Cells (HUtMEC).
Note: For best transfection results we recommend using PromoCell heparin-free Endothelial Cell Growth Medium MV2. Heparin, included all our other our Endothelial Cell Growth Media formulations, may reduce transfection efficiency.
Components: Part of a cell culture system offered in a variety of formats: Growth medium (ready-to-use): Includes basal medium and supplement mix growth medium kit: Includes basal medium and supplement pack supplement mix: Contains all media supplements premixed in one vial supplement pack: Contains all media supplements as individual vials.
Storage and stability: Store the basal medium at 4 to 8 °C in the dark immediately after arrival. Do not freeze the basal medium. If stored properly, the medium is stable until the expiry date stated on the label. After adding the supplements to the basal medium, the shelf life of the complete medium is 6 weeks at 4 to 8 °C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8 °C.