Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Spirotetramat Metabolite BYI08330-Cis-Keto-Hydroxy
Supelco® PESTANAL® Organic Reference Standard, Spirotetramat Metabolite BYI08330-Cis-Keto-Hydroxy
Catalog # 32485-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Spirotetramat Metabolite BYI08330-Cis-Keto-Hydroxy
Catalog # 32485-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    1172134-11-0
  • Description:
    Spirotetramat Metabolite BYI08330-cis-keto-hydroxy
  • Seq:
    03
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H317 - H319 - H335 - H361fd - H410
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    cis-3-(2,5-Dimethylphenyl)-3-hydroxy-8-methoxy-1-azaspiro[4.5]decane-2,4-dione
  • Formula:
    C₁₈H₂₃NO₄
  • Molecular weight:
    317,38
  • Storage temperature:
    −20 °C
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS09

Spirotetramat may be used as a reference standard for QuEChERS based methodology for the determination of spirotetramat and its relevant metabolites in pistachio samples by liquid chromatography-tandem mass spectrometry.

Caution: P202 - P273 - P280 - P302 + P352 - P305 + P351 + P338 - P308 + P313