Order Entry
Czech Republic
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Dextran From Leuconostoc Mesenteroides
Catalog # 31417-100MG-F
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Dextran From Leuconostoc Mesenteroides
Catalog # 31417-100MG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard, for GPC, Mw 5000
  • CAS No.:
    9004-54-0
  • Description:
    Dextran from Leuconostoc mesenteroides
  • Seq:
    11
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00130935
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Dextran Ladder
  • Formula:
    (C₆H₁₀O₅)ₙ
  • Flash point:
    Not applicable

Specifications

About this item