ContactUsLinkComponent
Portelite™ Fluorimetric Sodium Ion Quantification Kit
Portelite™ Fluorimetric Sodium Ion Quantification Kit
Catalog # AATB21327
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Portelite™ Fluorimetric Sodium Ion Quantification Kit
Catalog # AATB21327
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    50 Tests
  • Description:
    Portelite™ Fluorimetric Sodium Ion Quantification Kit
Specifications
About this item
Designed for consistent and dependable results. Portelite™ fluorimetric sodium ion quantification kit uses our robust sodium ion indicator dye, SoNa™ 520, which exhibit great fluorescence intensity enhancement upon binding to sodium ions.
  • Robust protocol
  • This nanodrop-based assay kit requires a small amount of sample, it is particularly suitable for the determination of sodium ion concentration in fields or on site
  • 12-month shelf life
SoNa™ 520 is perhaps the best sodium ion indicator for rapidly determining sodium ion concentration in combination with a fluorescence device such as a fluorescence Nanodrop spectrophotometer or a fluorescence microplate reader.
Delivery: Shipped at RT.
Caution: Research use only.