ContactUsLinkComponent
Amplite® Colorimetric Fructose-6-Phosphate Activity Assay Kit
Amplite® Colorimetric Fructose-6-Phosphate Activity Assay Kit
Catalog # AATB11330
Supplier:  AAT Bioquest
New Product
CAS Number:  
Amplite® Colorimetric Fructose-6-Phosphate Activity Assay Kit
Catalog # AATB11330
Supplier:  AAT Bioquest
New Product
CAS Number:  
Specifications
  • Pk:
    100 Tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® colorimetric fructose-6-phosphate assay kit is a highly sensitive and simple method of quantifying F6P present in a variety of samples.
  • Robust protocol
  • Absorbance measured at 450 nm
  • Six-month shelf life
One unit (U) is the amount of enzyme that catalyses the reaction of 1 µmol of substrate per minute.
Delivery: Shipped with blue ice.
Caution: Research use only.