ContactUsLinkComponent
Amplite® Colorimetric Phosphofructokinase (PFK) Activity Assay Kit
Amplite® Colorimetric Phosphofructokinase (PFK) Activity Assay Kit
Catalog # AATB11329
Supplier:  AAT Bioquest
New Product
CAS Number:  
Amplite® Colorimetric Phosphofructokinase (PFK) Activity Assay Kit
Catalog # AATB11329
Supplier:  AAT Bioquest
New Product
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Tests per kit:
    100 tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® Colorimetric Phosphofructokinase (PFK) Activity Assay Kit provides a simple and direct procedure for precise measurement of PFK enzyme activity in various biological samples.
  • Robust protocol
  • Absorbance measured at 450 nm
  • Six-month shelf life
One unit (U) is the amount of enzyme that catalyzes the reaction of 1 µmol of substrate per minute.
Delivery: Shipped with blue ice.
Caution: Research use only.