Order Entry
Czech Republic
ContactUsLinkComponent
Cumene
Flammable
Irritant
Health
Environment
Cumene
Catalog # CMPLSB224.500MG
New Product
CAS Number:  
Flammable
Irritant
Health
Environment
Cumene
Catalog # CMPLSB224.500MG
New Product
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
ADR:3,III
 :120.19 g/mol
UN:1918
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:98-82-8
Formula:C₉H₁₂
MDL Number:MFCD00008881