- Pk:100 mg
- CAS No.:122-34-9
- Description:Simazine
- Storage conditions:2...8 °C
- Hazard class description:H302 - H351 - H410
- Shipping conditions:STD
- MDL No.:MFCD00023174
- M-Clarity quality level:MQ300
- Synonym(s):2,4-Bis(ethylamino)-6-chloro-1,3,5-triazine
- Molecular weight:201,66
Simazine CRM may also be used as given below: Development and validation of an ultrasonic-assisted dispersive liquid-liquid microextraction (UA-DLLME) method for sample extraction followed by gas chromatography-mass spectrometry based determination of endocrine-disrupting herbicides in the river and seawater samples. Determination of triazine pesticide residues in samples of apples pears carrots potatoes tomatoes avocado cucumber spinach bananas and oranges by modified QuEChERS and solid-phase extraction (SPE) procedures followed by liquid chromatography-photodiode array (LC-DAD) analysis. Multi-residue analysis of 67 pesticides in olive oil olives and avocado samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS) to evaluate various clean-up sorbents. Quantitation of 20 pesticides in eel and shrimp samples by a scheduled multiple reaction monitoring (sMRM) method and QuEChERS with EDTA for sample extraction and purification. Analysis of 35 micropollutants including pesticides in water samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS) after their solid-phase extraction. Simultaneous quantification of 58 pharmaceuticals and personal care products (PPCPs) and pesticides in water samples by solvent-based de-emulsification dispersive liquid-liquid microextraction (SD-DLLME) combined with liquid chromatography-tandem mass spectrometry (LCMS/MS).