Supelco® Organic Reference Standard, F.A.M.E. Mix RM-6
Catalog # O7631-1AMP
Supplier: Merck
CAS Number:
Specifications
- Concentration:Pk of 100 mg (Neat)
- Pk:1 Ampoul
- Standards:Certified reference material
- Description:F.A.M.E. mix RM-6
- Storage conditions:–20 °C
- Shipping conditions:DICE
- M-Clarity quality level:MQ100
- Synonym(s):AOCS Reference Mixtures; AOCS No. 6 (RM-6); Oil Reference Standard, AOCS No. 6; Oil Reference Standard, AOCS
Specifications
About this item
Myristic acid methyl ester (C14:0) 2,0% Palmitic acid methyl ester (C16:0) 30,0% Palmitoleic acid methyl ester (C16:1cis-9) 3,0% Stearic acid methyl ester (C18:0) 14,0% Oleic acid methyl ester (C18:1cis-9) 41,0% Linoleic acid methyl ester (C18:2cis-912) 7,0% Linolenic acid methyl ester (C18:3cis-91215) 3,0%.
- GHS signal: GHS05