Supelco® Organic Reference Standard, Pullulan Standard Set Mp ~350 - 700′000
96351-1KT
: Merck
:
- Pk:1 KIT
- Standards:Set of analytical standards, for GPC
- CAS No.:9057-02-7
- Description:Pullulan standard set Mp ~350 - 700′000
- Seq:30
- Storage conditions:2…8 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):Pullulan Standard Set