Supelco® Cerilliant® Organic Reference Standard, Acylcarnitines Mix 1 (C0,C2) hydrochloride solution
Katalogové číslo A-141-1ML
Dodavatel: Merck
Číslo CAS:
Specifikace
- Concentration:10 mg/ml in methanol (each analyte as free base)
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- Description:Acylcarnitines mix 1 (C0,C2) hydrochloride solution
- Storage conditions:–70 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:WICE
- M-Clarity quality level:MQ300
- Molecular weight:197,66
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
Specifikace
O tomto produktu
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311