Supelco® Organic Reference Standard, Bacoside A, mixture of Bacoside A3, Bacopaside II, Bacopaside X and Bacopasaponin C
Catalog # 76091-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:Analytical standard
- Description:Bacoside A, mixture of Bacoside A3, Bacopaside II, Bacopaside X and Bacopasaponin C
- Storage conditions:2...30 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100