Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Chlordane (mixture of isomers)
Supelco® TraceCERT® Organic Reference Standard, Chlordane (mixture of isomers)
Catalog # 53797-20MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, Chlordane (mixture of isomers)
Catalog # 53797-20MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    20 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    57-74-9
  • Description:
    Chlordane (mixture of isomers)
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H301 + H311 - H351 - H410
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ300

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P202 - P264 - P273 - P280 - P301 + P310 - P302 + P352 + P312