Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Protonex™ Red 780 AM
Protonex™ Red 780 AM cell permeable
Protonex™ Red 780 AM cell permeable
Protonex™ Red 780 AM cell permeable
Katalogové číslo AATB21186
Dodavatel:
AAT Bioquest
New Product
Číslo CAS:
Rozbalit obrázek
Protonex™ Red 780 AM cell permeable
Katalogové číslo AATB21186
Dodavatel:
AAT Bioquest
New Product
Číslo CAS:
Specifikace
Pk:
1 mg
Seq:
2070
Specifikace
O tomto produktu
Detaily produktu
:
:
798.89 g/mol
Detaily produktu