Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
iFluor® 430 tyramide
iFluor® 430 tyramide [Superior Replacement for Opal 480]
iFluor® 430 tyramide [Superior Replacement for Opal 480]
iFluor® 430 tyramide [Superior Replacement for Opal 480]
Catalog # AATB45096
Supplier:
AAT Bioquest
New Product
CAS Number:
Expand image
iFluor® 430 tyramide [Superior Replacement for Opal 480]
Catalog # AATB45096
Supplier:
AAT Bioquest
New Product
CAS Number:
Specifications
Pk:
200
Seq:
0200
Specifications
About this item
Product Details
:
:
710.89 g/mol
Product Details