Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Helixyte™ 14
Helixyte™ 14 5 mM [Equivalent to SYBR 14]
Helixyte™ 14 5 mM [Equivalent to SYBR 14]
Helixyte™ 14 5 mM [Equivalent to SYBR 14]
Catalog # AATB17563
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
Helixyte™ 14 5 mM [Equivalent to SYBR 14]
Catalog # AATB17563
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
0,5 mL
Seq:
2040
Specifications
About this item
Product Details
:
:
474.06 g/mol
Product Details