Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bodi Fluor FL-C12
Bodi Fluor FL-C12 [equivalent to BODIPY™ FL C12]
Bodi Fluor FL-C12 [equivalent to BODIPY™ FL C12]
AATB22121
:
AAT BIOQUEST
New Product
:
Bodi Fluor FL-C12 [equivalent to BODIPY™ FL C12]
AATB22121
:
AAT BIOQUEST
New Product
:
Pk:
1 mg
Seq:
2070
:
:
418.34 g/mol