Bio-16-UTP [Biotin-16-UTP] 1 mM
New Product
:
:963.78 g/mol
Frequently Bought Together
Transfer pipettes
: RATIOLAB
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Ice Pans, with or without Lid
: VWR Collection
Deep well storage plates, 96-well, 0,8 ml
: THERMO MATRIX TECHNOLOGIES
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
: VWR Collection
VWR®, Test Tubes
: VWR Collection
VWR® Premium Line, Serological Pipettes
: VWR Collection
Cell strainers, Corning®
: Corning
Cryogenic vials, Cryovial®
: Simport Scientific
Cryoboxes, 136×136 mm
: RATIOLAB
Cell culture flasks, Nunc™
: Thermo Fisher Scientific
VWR®, Flat Screw Caps for Microcentrifuge Tubes
: VWR Collection
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
: VWR Collection
Cell culture flasks, Falcon®
: Corning
VWR®, Cryoboxes, Cardboard, 136x136 mm
: VWR Collection
Accessories and consumables for KingFisher™ Flex
: Thermo Fisher Scientific