Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(and-6)-Carboxy SNARF-4F AM ester
5-(and-6)-Carboxy SNARF-4F AM ester
5-(and-6)-Carboxy SNARF-4F AM ester
5-(and-6)-Carboxy SNARF-4F AM ester
Catalog # AATB21173
Supplier:
AAT Bioquest
New Product
CAS Number:
Expand image
5-(and-6)-Carboxy SNARF-4F AM ester
Catalog # AATB21173
Supplier:
AAT Bioquest
New Product
CAS Number:
Specifications
Pk:
50 µG
Seq:
1202
Pk Info:
10 x 50 µg
Specifications
About this item
Product Details
:
:
585.54 g/mol
Product Details