Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(and-6)-Carboxy SNARF-4F
5-(and-6)-Carboxy SNARF-4F
5-(and-6)-Carboxy SNARF-4F
5-(and-6)-Carboxy SNARF-4F
AATB21170
:
AAT Bioquest
New Product
:
5-(and-6)-Carboxy SNARF-4F
AATB21170
:
AAT Bioquest
New Product
:
Pk:
1 mg
Seq:
2070
:
:
471.44 g/mol