Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HIF-2-alpha Rabbit Polyclonal Antibody
Anti-HIF-2-alpha Rabbit Polyclonal Antibody
Catalog # ANTIA10163-100
Supplier:  Antibodies.com
New Product
undefined
Anti-HIF-2-alpha Rabbit Polyclonal Antibody
Catalog # ANTIA10163-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Basic helix loop helix PAS protein MOP2
  • Antigen symbol:
    HIF-2-alpha
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q99814
  • Antigen synonyms:
    bHLHe73|Basic-helix-loop-helix-PAS protein MOP2|Endothelial pas domain protein 1|ECYT4|Class E basic helix-loop-helix protein 73|EPAS-1|EPAS 1|Endothelial PAS domain containing protein 1|Endothelial PAS domain-containing protein 1
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,05% Proclin 300
  • Molecular weight:
    120 kDa
  • Sequence:
    TFKTRSAKGFGARGPDVLSPAMVALSNKLKLKRQLEYEEQAFQDLSGGDPPGGSTSHLMWKRMKNLRG
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 678 - 745 of human EPAS1/HIF2a (NP_001421.2)
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to HIF-2 alpha for WB, IHC and ICC/IF with samples derived from human and mouse.

  • Validated Applications: WB, IHC, ICC/IF
  • Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: HIF-2-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse