Order Entry
Czech Republic
ContactUsLinkComponent
Anti-KDM4B/JMJD2B Rabbit Polyclonal Antibody
Anti-KDM4B/JMJD2B Rabbit Polyclonal Antibody
  ANTIA100665-100
New Product
undefined
Anti-KDM4B/JMJD2B Rabbit Polyclonal Antibody
  ANTIA100665-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    JHDM3B
  • Antigen symbol:
    KDM4B/JMJD2B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O94953
  • Antigen synonyms:
    Kdm4b|KIAA0876|JmjC domain-containing histone demethylation protein 3B|KDM4B_HUMAN|JMJD2B|KDM4B / JMJD2B|Jumonji domain containing 2B|Jumonji domain-containing protein 2B|Jumonji domain containing protein 2B
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    121 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human KDM4B (amino acids 351 - 400)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody to JHD3B for IHC and ELISA with samples derived from human.

  • Validated Applications: IHC, ELISA
  • Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:1000

Type: Primary
Antigen: KDM4B / JMJD2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human