Anti-SLC6A15 Rabbit Polyclonal Antibody
Catalog # ANTIA100092-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:hv7 3
- Antigen symbol:SLC6A15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9H2J7
- Antigen synonyms:NTT73|Sodium-coupled branched-chain amino-acid transporter 1|SLC6A15|S6A15_HUMAN|Orphan sodium and chloride dependent neurotransmitter transporter NTT73|Orphan transporter v7 3|SBAT1|sodium- and chloride-dependent neurotransmitter transporter NTT73|MGC87066
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:81 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human SLC6A15 (amino acids 32 - 81)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to SLC6A15 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: SLC6A15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human