Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PVRL4 [EPR15613-68] Rabbit Polyclonal Antibody
Anti-PVRL4 [EPR15613-68] Rabbit Polyclonal Antibody
Catalog # ANTIA100796-100
Supplier:  Antibodies.com
New Product
undefined
Anti-PVRL4 [EPR15613-68] Rabbit Polyclonal Antibody
Catalog # ANTIA100796-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DKFZp686K05193
  • Antigen symbol:
    PVRL4 [EPR15613-68]
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q96NY8
  • Antigen synonyms:
    EDSS1|Nectin-4|Processed poliovirus receptor-related protein 4|Nectin 4|PRR4|Ig superfamily receptor LNIR|poliovirus receptor-related 4|pvrl4|PVRL4 [EPR15613-68]
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    55 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human PVRL4 (amino acids 312-361)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to PVRL4 for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000

Type: Primary
Antigen: PVRL4 [EPR15613-68]
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human