Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLC39A14 / ZIP-14 Rabbit Polyclonal Antibody
Anti-SLC39A14 / ZIP-14 Rabbit Polyclonal Antibody
  ANTIA8608-100
New Product
undefined
Anti-SLC39A14 / ZIP-14 Rabbit Polyclonal Antibody
  ANTIA8608-100
New Product

 

  • Antibody type:
    Primary
  • Antigen symbol:
    SLC39A14 / ZIP-14
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q15043
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,05% Proclin 300
  • Molecular weight:
    54 kDa
  • Sequence:
    SSLGAPAISAASFLQDLIHRYGEGDSLTLQQLKALLNHLDVGVGRGNVTQHVQGHRNLSTCFSSGDLFTAHNFSEQSRIGSSELQEFCPTILQQLDSRACTSENQENEENEQTEEGRPSAVEVWGYG
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 31-157 of human ZIP14 (NP_001121903.1)
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to SLC39A14 / ZIP-14 for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, IHC, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:20

Type: Primary
Antigen: SLC39A14 / ZIP-14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat