Anti-CD264 Mouse Monoclonal Antibody [Clone: TRAIL-R4-01]
ANTIA86722-100
New Product
- Antibody type:Primary
- Antigen name:CD 264
- Antigen symbol:CD264
- Clonality:Monoclonal
- Clone:TRAIL-R4-01
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q9UBN6
- Antigen synonyms:TNF receptor related receptor for TRAIL|TNF related apoptosis inducing ligand receptor 4|DcR 2|Decoy with truncated death domain|DcR2|OTTHUMP00000123528|CD264 antigen|CD264|Decoy receptor 2
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,4, without Sodium Azide (0,2 µm filter sterilised)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:TRAIL-R4 (aa 1-210) - hIgGhc fusion protein
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [TRAIL-R4-01] antibody to CD264 for Flow Cytometry with samples derived from Human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: CD264
Clonality: Monoclonal
Clone: TRAIL-R4-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human