Anti-HLA B7 Mouse Monoclonal Antibody [Clone: BB7.1]
Katalogové číslo ANTIA86658-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Abacavir hypersensitivity, susceptibility to
- Antigen symbol:HLA B7
- Clonality:Monoclonal
- Clone:BB7.1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Non-human primates,Human
- Form:Liquid
- Antigen synonyms:HLA class I histocompatibility antigen B 7 alpha chain|HLA class I histocompatibility antigen, B-15 alpha chain|HLA class I histocompatibility antigen, B-18 alpha chain|HLA B|HLA class I histocompatibility antigen, B-13 alpha chain|HLA class I histocompatibility antigen, B-27 alpha chain|Drug-induced liver injury due to flucloxacillin|HLA B7|HLA class I histocompatibility antigen, B-14 alpha chain
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Papain solubilised HLA-A2, B7
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [BB7.1] antibody to HLA B7 for Flow Cytometry with samples derived from Human and Non-Human Primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: HLA B7
Clonality: Monoclonal
Clone: BB7.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates