Anti-CD58 Mouse Monoclonal Antibody [Clone: MEM-63] (Biotinylated)
Catalog # ANTIA86371-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:AG3
- Antigen symbol:CD58
- Clonality:Monoclonal
- Clone:MEM-63
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Pig,Human
- Form:Liquid
- Gene ID:UniprotID# P19256
- Antigen synonyms:LFA-3|CD58 antigen|CD58 molecule|CD58 antigen, (lymphocyte function associated antigen 3)|CD58 antigen (lymphocyte function associated antigen 3)1|LFA 3|FLJ23181|FLJ43722|CD58
- Storage buffer:Supplied in Tris Buffered Saline, pH 8,0, with 15 mM Sodium Azide
- Storage temperature:Store at 2...8 °C. Do not freeze!
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:NALM-6 human pre-B cell line
- Purification:This antibody is conjugated with Biotin-LC-NHS under optimum conditions. The reagent is free of unconjugated Biotin.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [MEM-63] antibody to CD58 (Biotin) for Flow Cytometry with samples derived from Human and Porcine.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 2 to 12 µg/ml
Type: Primary
Antigen: CD58
Clonality: Monoclonal
Clone: MEM-63
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Porcine