Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD58 Mouse Monoclonal Antibody [Clone: MEM-63] (FITC (Fluorescein Isothiocyanate))
Anti-CD58 Mouse Monoclonal Antibody [Clone: MEM-63] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA86369-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD58 Mouse Monoclonal Antibody [Clone: MEM-63] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA86369-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AG3
  • Antigen symbol:
    CD58
  • Clonality:
    Monoclonal
  • Clone:
    MEM-63
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P19256
  • Antigen synonyms:
    LFA-3|CD58 antigen|CD58 molecule|CD58 antigen, (lymphocyte function associated antigen 3)|CD58 antigen (lymphocyte function associated antigen 3)1|LFA 3|FLJ23181|FLJ43722|CD58
  • Storage buffer:
    Supplied in Tris Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    NALM-6 human pre-B cell line
  • Purification:
    This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [MEM-63] antibody to CD58 (FITC) for Flow Cytometry with samples derived from Human and Porcine.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD58
Clonality: Monoclonal
Clone: MEM-63
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Porcine