Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD3 Mouse Monoclonal Antibody [Clone: MEM-57] (PerCP (Peridinin-Chlorophyll Protein Complex))
Anti-CD3 Mouse Monoclonal Antibody [Clone: MEM-57] (PerCP (Peridinin-Chlorophyll Protein Complex))
Catalog # ANTIA86064-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD3 Mouse Monoclonal Antibody [Clone: MEM-57] (PerCP (Peridinin-Chlorophyll Protein Complex))
Catalog # ANTIA86064-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    4930549J05Rik
  • Antigen symbol:
    CD3
  • Clonality:
    Monoclonal
  • Clone:
    MEM-57
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P07766
  • Antigen synonyms:
    CD247 antigen|CD3|CD3 antigen, delta subunit|CD247 antigen, zeta subunit|CD247 molecule|Cd247|CD3 delta|A430104F18Rik|AW552088
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human thymocytes and T lymphocytes
  • Purification:
    This antibody is conjugated with PerCP under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [MEM-57] antibody to CD3 (PerCP) for Flow Cytometry with samples derived from Human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: MEM-57
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human