Anti-HLA E Mouse Monoclonal Antibody [Clone: MEM-E/07]
Catalog # ANTIA86034-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:EA1.2
- Antigen symbol:HLA E
- Clonality:Monoclonal
- Clone:MEM-E/07
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P13747
- Antigen synonyms:EA2.1|HLA class I histocompatibility antigen E alpha chain precursor|HLA E|HLA 6.2|HLA6.2|Lymphocyte antigen|HLAE|HLA class I histocompatibility antigen alpha chain E|HLA class I histocompatibility antigen E alpha chain
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Bacterially expressed recombinant HLA-E refolded with beta2-microglobulin and peptide
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [MEM-E/07] antibody to HLA E for Flow Cytometry, IP and IHC-P with samples derived from Human.
- Validated applications: Flow cytometry, IP, IHC-P
- Recommended dilutions: IHC-P: 10 µg/ml, Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: HLA E
Clonality: Monoclonal
Clone: MEM-E/07
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human